missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™

46.70 € - 200.00 €

Chemical Identifiers

CAS 375857-64-0
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
InChI Key NYAFKUCVAVUCHW-UHFFFAOYSA-N
PubChem CID 18001310
IUPAC Name 5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
SMILES CC1=NOC(=N1)C2=CC=CC=C2Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10015003
SDS View Documents
Thermo Scientific Maybridge
CC49610CB
250 mg
46.70 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10788635
SDS View Documents
Thermo Scientific Maybridge
CC49610DA
1 g
59.60 €
1g
Please to purchase this item. Need a web account? Register with us today!
10178794
SDS View Documents
Thermo Scientific Maybridge
CC49610DE
5 g
200.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

375857-64-0
239.072
18001310
CC1=NOC(=N1)C2=CC=CC=C2Br
C9H7BrN2O
NYAFKUCVAVUCHW-UHFFFAOYSA-N
5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole

Specifications

375857-64-0
C9H7BrN2O
NYAFKUCVAVUCHW-UHFFFAOYSA-N
5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
18001310
97%
97%
5-2-bromophenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-2-bromophenyl-3-methyl, 5-2-bromophenyl-3-methyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-2-bromophenyl-3-methyl
CC1=NOC(=N1)C2=CC=CC=C2Br
239.072
239.07
5-(2-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.