missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Chloro-4-nitrophenyl)-2-furoyl chloride, Thermo Scientific™

87.00 € - 385.00 €

Chemical Identifiers

CAS 380594-11-6
Molecular Formula C11H5Cl2NO4
Molecular Weight (g/mol) 286.064
MDL Number MFCD02258011
InChI Key BQTYBZIOLAXUSL-UHFFFAOYSA-N
Synonym 5-2-chloro-4-nitrophenyl furan-2-carbonyl chloride, 5-2-chloro-4-nitrophenyl-2-furoyl chloride, 5-2-chloro-4-nitro-phenyl-furan-2-carbonyl chloride, acmc-1ctg9, 5-2-chloro-4-nitrophenyl-2-furoylchloride
PubChem CID 4484926
IUPAC Name 5-(2-chloro-4-nitrophenyl)furan-2-carbonyl chloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)C2=CC=C(O2)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355399
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50400.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11365399
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50400.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

380594-11-6
286.064
BQTYBZIOLAXUSL-UHFFFAOYSA-N
4484926
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C2=CC=C(O2)C(=O)Cl
C11H5Cl2NO4
MFCD02258011
5-2-chloro-4-nitrophenyl furan-2-carbonyl chloride, 5-2-chloro-4-nitrophenyl-2-furoyl chloride, 5-2-chloro-4-nitro-phenyl-furan-2-carbonyl chloride, acmc-1ctg9, 5-2-chloro-4-nitrophenyl-2-furoylchloride
5-(2-chloro-4-nitrophenyl)furan-2-carbonyl chloride

Specifications

380594-11-6
MFCD02258011
5-2-chloro-4-nitrophenyl furan-2-carbonyl chloride, 5-2-chloro-4-nitrophenyl-2-furoyl chloride, 5-2-chloro-4-nitro-phenyl-furan-2-carbonyl chloride, acmc-1ctg9, 5-2-chloro-4-nitrophenyl-2-furoylchloride
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C2=CC=C(O2)C(=O)Cl
286.064
286.07
C11H5Cl2NO4
UN3261
BQTYBZIOLAXUSL-UHFFFAOYSA-N
5-(2-chloro-4-nitrophenyl)furan-2-carbonyl chloride
4484926
5-(2-Chloro-4-nitrophenyl)-2-furoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications