missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)-1-methyl-1H-pyrazole-3-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. p-3175287
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10679934

Brand: Thermo Scientific Maybridge CC46901CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 108128-39-8
Molecular Formula C9H8N2O3
Molecular Weight (g/mol) 192.17
MDL Number MFCD08060535
InChI Key XXEWHNSDJMBLBK-UHFFFAOYSA-N
Synonym 5-2-furyl-1-methyl-1h-pyrazole-3-carboxylic acid, 5-furan-2-yl-1-methylpyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl-1-methyl, 5-furan-2-yl-1-methyl-1h-pyrazole-3-carboxylic acid, acmc-1btkv, 5-2-furyl-1-methylpyrazole-3-carboxylic acid
PubChem CID 7537639
SMILES CN1N=C(C=C1C1=CC=CO1)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 5-(furan-2-yl)-1-methylpyrazole-3-carboxylic acid
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 5-(2-Furyl)-1-methyl-1H-pyrazole-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.