missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10761114
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 400715-45-9
Molecular Formula C9H7NO2S2
Molecular Weight (g/mol) 225.28
MDL Number MFCD02682053
InChI Key AEDWMWQOFJYAGN-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-carboxylic acid, 5-2-methylthiazol-4-yl-thiophene-2-carboxylic acid, 5-2-methylthiazol4-yl-thiophene-2-carboxylic acid, thiophene-2-carboxylic acid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid #
PubChem CID 614467
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
SMILES CC1=NC(=CS1)C2=CC=C(S2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 225.29
Percent Purity 97%
Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)-2-thiophenecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'