missing translation for 'onlineSavingsMsg'
Learn More

5-[2-Methyl-3-(trifluoromethyl)-1H-2lambda∼5∼-pyrazol-5-yl]-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10337304
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 223499-20-5
Molecular Formula C10H7F3N2O2S
Molecular Weight (g/mol) 276.233
MDL Number MFCD00206674
InChI Key MGYVPTDPOYVOQK-UHFFFAOYSA-N
Synonym 5-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-2-carboxylic acid, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl thiophene-2-carboxylic acid, 3-5-carboxythien-2-yl-1-methyl-5-trifluoromethyl-1h-pyrazole, 2-thiophenecarboxylicacid, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-thiophene-2-carboxylic acid, 5-1-methyl-5-trifluoromethyl-1h-pyrazole-3-yl-2-thiophenecarboxylic acid
PubChem CID 2775669
IUPAC Name 5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carboxylic acid
SMILES CN1C(=CC(=N1)C2=CC=C(S2)C(=O)O)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 276.24
Percent Purity 97%
Chemical Name or Material 5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'