missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Thienyl)isoxazole, 97%, Thermo Scientific™

Product Code. 15436037
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138716-44-6
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.18
MDL Number MFCD06409150
InChI Key KNRAAHWCKJMGNF-UHFFFAOYSA-N
Synonym 5-2-thienyl isoxazole, 5-thiophen-2-yl-isoxazole, 5-thiophen-2-yl isoxazole, 5-thiophen-2-yl-1,2-oxazole, 5-thiophene-2-yl-isoxazole, 5-2-thiophenyl isoxazole, 5-2-thienyl-isoxazole, isoxazole, 5-2-thienyl, isoxazole, 5-2-thienyl-9ci
PubChem CID 12049986
IUPAC Name 5-thiophen-2-yl-1,2-oxazole
SMILES O1N=CC=C1C1=CC=CS1

Specifications

Boiling Point 68°C to 70°C (0.22 mmHg)
Refractive Index 1.613
Quantity 10 g
Formula Weight 151.19
Percent Purity 97%
Chemical Name or Material 5-(2-Thienyl)isoxazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.