missing translation for 'onlineSavingsMsg'
Learn More
5-(3-Pyridinyloxy)-2-furoic acid, ≥97%, Thermo Scientific™ Product Code.: 10472903

5-(3-Pyridinyloxy)-2-furoic acid, ≥97%, Thermo Scientific™

Product Code. 10472903
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10472903

Brand: Thermo Scientific Maybridge AW01246DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-39-3
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.169
MDL Number MFCD03407385
InChI Key ZTYMSWYWSCYFAF-UHFFFAOYSA-N
Synonym 5-3-pyridinyloxy-2-furoic acid, 5-pyridin-3-yloxy furan-2-carboxylic acid, 2-furancarboxylic acid,5-3-pyridinyloxy, 5-3-pyridyloxy furan-2-carboxylic acid, 5-3-pyridinyl-oxy-2-furoic acid, 5-pyridin-3-yl oxy furan-2-carboxylic acidShow MoreShow Less
PubChem CID 7060545
IUPAC Name 5-pyridin-3-yloxyfuran-2-carboxylic acid
SMILES C1=CC(=CN=C1)OC2=CC=C(O2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.17
Percent Purity 97%
Chemical Name or Material 5-(3-Pyridinyloxy)-2-furoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(3-Pyridinyloxy)-2-furoic acid, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.