missing translation for 'onlineSavingsMsg'
Learn More
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan, 97%, Thermo Scientific Chemicals Product Code.: 10754502

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan, 97%, Thermo Scientific Chemicals

Product Code. 10754502
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10754502

Brand: Thermo Scientific Acros 430470050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073355-14-2
Molecular Formula C12H15BN2O3
Molecular Weight (g/mol) 246.07
MDL Number MFCD09842717
InChI Key MQXANAGMQAYTMV-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo c 1,2,5 oxadiazole, benzofurazan-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzoxadiazole, benzofurazan-5-boronic acid, pinacol ester, benzo c 1,2,5 oxadiazole-5-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzofurazan, benzo c 1,2,5 oxadiazole-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzoxadiazole, benzo c 1,2,5 oxadiazol-5-ylboronic acid pinacol ester, amtb001Show MoreShow Less
PubChem CID 46738004
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzoxadiazole
SMILES CC1(C)OB(OC1(C)C)C1=CC2=NON=C2C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 88.0°C to 90.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 246.08
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number339759 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number430470010,430470050
Lot Number339759 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number430470010,430470050
compliance-icons
Product Identifier
  • 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.