missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Fluorophenyl)isoxazole, 99%, Thermo Scientific™

Product Code. 11356589
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138716-37-7
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.151
MDL Number MFCD02183544
InChI Key JZCBQTRPILCIBC-UHFFFAOYSA-N
Synonym 5-4-fluorophenyl isoxazole, 5-4-fluorophenyl-1,2-oxazole, acmc-1c1fl, 1-fluoro-4-isoxazol-5-yl benzene, 1-fluoro-4-1,2-oxazol-5-yl benzene
PubChem CID 2774719
IUPAC Name 5-(4-fluorophenyl)-1,2-oxazole
SMILES C1=CC(=CC=C1C2=CC=NO2)F

Specifications

Melting Point 53°C to 55°C
Quantity 5 g
Formula Weight 163.15
Percent Purity 99%
Chemical Name or Material 5-(4-Fluorophenyl)isoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'