missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethyl-1,3-cyclohexanedione, 99%, Thermo Scientific Chemicals

Product Code. 10666492
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-81-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001588
InChI Key BADXJIPKFRBFOT-UHFFFAOYSA-N
Synonym dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
PubChem CID 31358
IUPAC Name 5,5-dimethylcyclohexane-1,3-dione
SMILES CC1(C)CC(=O)CC(=O)C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 147.0°C to 150.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2C6H6(=O)2
Quantity 25 g
Beilstein 07, 559
Fieser 01,266
Merck Index 15, 3264
Solubility Information Solubility in water: 5 % (25°C). Other solubilities: soluble in methanol,ethanol,benzene,chloroform,and acetic acid
Formula Weight 140.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 5, 5-Dimethyl-1, 3-cyclohexanedione, 99%
Show More Show Less
Product Identifier
  • 5,5-Dimethyl-1,3-cyclohexanedione

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'