missing translation for 'onlineSavingsMsg'
Learn More
5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid, 97%, Thermo Scientific™ Product Code.: 10475322

5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid, 97%, Thermo Scientific™

Product Code. 10475322
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10475322

Brand: Thermo Scientific Maybridge KM06169DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 238742-86-4
Molecular Formula C12H8F3NO3S
Molecular Weight (g/mol) 303.26
MDL Number MFCD00099971
InChI Key MYDIGMNTURCKBI-UHFFFAOYSA-N
Synonym 5-5-trifluoromethyl-2-pyridyl thio methyl-2-furoic acid, 5-5-trifluoromethyl pyridin-2-yl sulfanyl methyl furan-2-carboxylic acid, 2-5-carboxyfurfurylthio-5-trifluoromethyl pyridine, 5-5-trifluoromethyl pyridin-2-yl thiomethyl-2-furoic acid, maybridge1_004760, 5-5-trifluoromethyl-2-pyridyl thiomethyl-2-furoic acid, 2-furancarboxylicacid, 5-5-trifluoromethyl-2-pyridinyl thio methyl, 5-5-trifluoromethyl pyridin-2-yl thio methyl furan-2-carboxylic acid, 5-5-trifluoromethyl pyridin-2-yl sulfanylmethyl furan-2-carboxylic acidShow MoreShow Less
PubChem CID 2736458
IUPAC Name 5-[[5-(trifluoromethyl)pyridin-2-yl]sulfanylmethyl]furan-2-carboxylic acid
SMILES OC(=O)C1=CC=C(CSC2=NC=C(C=C2)C(F)(F)F)O1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 303.26
Percent Purity 97%
Chemical Name or Material 5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.