missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ 5(6)-CFDA, SE; CFSE (5-(and-6)-Carboxyfluorescein Diacetate, Succinimidyl Ester), mixed isomers

5(6)-CFDA, SE; CFSE (5-(and-6)-Carboxyfluorescein Diacetate, Succinimidyl Ester), mixed isomers

Brand:  Molecular Probes™ C1157

451.25 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 11524217

  • 490.00 € / 25mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CFDA, SE is a fixable-cell-permeant, fluorescein-based tracer for very long-term cell labeling. The dye is also available in our convenient, easy-to-use Vybrant CFDA-SE Cell Tracer Kit (V12883).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 1 vial of CFSE (25 mg). Store in freezer (-5 to -30°C).
CFDA SE (CFSE)
25 mg
Solid
DMSO (Dimethylsulfoxide)
Tracer
492⁄517
Fluorescence Microscope, Flow Cytometer
Fluorescence
Room Temperature
Classic Dyes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.