missing translation for 'onlineSavingsMsg'
Learn More
5,6-Dichloro-2-mercaptobenzimidazole, 98%, Thermo Scientific Chemicals Product Code.: 11436847

5,6-Dichloro-2-mercaptobenzimidazole, 98%, Thermo Scientific Chemicals

Product Code. 11436847
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11436847

Brand: Thermo Scientific Alfa Aesar A14309.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed in the synthesis of 2?,3?-dideoxy and 2?,3?-unsaturated ribofuranonucleosides as potential antiviral agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of 2′,3′-dideoxy and 2′,3′-unsaturated ribofuranonucleosides as potential antiviral agents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19462-98-7
Molecular Formula C7H4Cl2N2S
Molecular Weight (g/mol) 219.083
MDL Number MFCD00052398
InChI Key AFDOMGKBKBKUHB-UHFFFAOYSA-N
PubChem CID 2774259
IUPAC Name 5,6-dichloro-1,3-dihydrobenzimidazole-2-thione
SMILES C1=C2C(=CC(=C1Cl)Cl)NC(=S)N2

Specifications

Melting Point ∼350°C (decomposition)
Odor Odorless
Quantity 1 g
Synonym 5,6-dichloro-1h-benzo d imidazole-2-thiol, 5,6-dichlorobenzimidazole-2-thiol, 5,7dicl2sh-bzind, 2-mercapto-5,6-dichlorobenzimidazole, 5,6-dichloro-1h-benzimidazole-2-thiol, 5,6-dichloro-2-mercaptobenzimidazole, 5,6-dichloro-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione,5,6-dichloro-1,3-dihydro, 2h-benzimidazole-2-thione, 5,6-dichloro-1,3-dihydro, 5,6-dichloro-1,3-dihydro-1,3-benzodiazole-2-thione
Solubility Information Insoluble in water.
Formula Weight 219.09
Percent Purity 98%
Chemical Name or Material 5,6-Dichloro-2-mercaptobenzimidazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ1590A Certificate TypeCertificate of Analysis Date19/01/2023 Catalog NumberA14309.03,A14309.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,6-Dichloro-2-mercaptobenzimidazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.