missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Product Code. 11319819
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3002-81-1
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD00004983
InChI Key BRPQDJPJBCQFSR-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 5,6-dimethyl, acmc-1afjl, brpqdjpjbcqfsr-uhfffaoysa, 5,6-dimethylpyridino 3,2-h quinoline, 5,6-dimethyl-1,10-phenanthroline
PubChem CID 76357
IUPAC Name 5,6-dimethyl-1,10-phenanthroline
SMILES CC1=C2C=CC=NC2=C3C(=C1C)C=CC=N3

Specifications

Melting Point 266°C to 269°C
Quantity 1 g
Beilstein 160918
Sensitivity Hygroscopic
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material 5,6-Dimethyl-1,10-phenanthroline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'