missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylthiophene-2-boronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11328216
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206551-43-1
Molecular Formula C6H7BO3S
Molecular Weight (g/mol) 169.989
MDL Number MFCD01075681
InChI Key DCNMATSPQKWETQ-UHFFFAOYSA-N
Synonym 5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
PubChem CID 4143377
IUPAC Name (5-acetylthiophen-2-yl)boronic acid
SMILES B(C1=CC=C(S1)C(=O)C)(O)O

Specifications

Melting Point 142°C to 148°C
Quantity 1 g
Beilstein 8681302
Formula Weight 170
Percent Purity 98%
Chemical Name or Material 5-Acetylthiophene-2-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'