missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Acetylthiophene-2-carbonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 151.19 |
Percent Purity | 97% |
Chemical Name or Material | 5-Acetylthiophene-2-carbonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'