missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyluracil, 98%, Thermo Scientific™

Product Code. 11408560
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6214-65-9
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00151968
InChI Key YNYDWEIQSDFDLK-UHFFFAOYSA-N
Synonym 5-acetyluracil, 5-acetylpyrimidine-2,4 1h,3h-dione, 1-2,4-dihydroxypyrimidin-5-yl ethanone, 2,4 1h,3h-pyrimidinedione, 5-acetyl, 5-acetyl-1,3-dihydropyrimidine-2,4-dione, 5-acetyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 5-acetyl-2,6-dioxo-1,3-dihydropyrimidine, 5-acetyl uracil, acetylpyrimidinedione, zlchem 449
PubChem CID 234581
SMILES CC(=O)C1=CNC(=O)NC1=O

Specifications

Quantity 1 g
IUPAC Name 5-acetyl-1H-pyrimidine-2,4-dione
Formula Weight 154.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Acetyluracil
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'