missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-methyl-1H-indazole, 97%, Thermo Scientific™

Product Code. 10309747
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 50593-24-3
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD03305455
InChI Key PYOFNPHTKBSXOM-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazol-5-amine, 5-amino-1-methyl-1h-indazole, 1-methyl-1h-indazol-5-ylamine, 1h-indazol-5-amine, 1-methyl, 5-amino-1-methylindazole, 1-methyl-1h-indazole-5-amine, pubchem20587, 1-methyl-5-aminoindazol, 1-methyl-5-indazolamine, 1-methyl-5-aminoindazole
PubChem CID 2768032
IUPAC Name 1-methylindazol-5-amine
SMILES CN1C2=C(C=C(C=C2)N)C=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 154.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 147.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1-methyl-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-1-methyl-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'