Learn More
5-Amino-2,3-dihydrobenzo[b]furan, 97%, Thermo Scientific Chemicals
57.70 € - 479.00 €
Chemical Identifiers
CAS | 42933-43-7 |
---|---|
Molecular Formula | C8H9NO |
Molecular Weight (g/mol) | 135.166 |
MDL Number | MFCD03617969 |
InChI Key | YJMADHMYUJFMQE-UHFFFAOYSA-N |
Synonym | 2,3-dihydrobenzofuran-5-amine, 2,3-dihydrobenzo b furan-5-ylamine, 5-amino-2,3-dihydrobenzo b furan, 5-aminocoumaran, 5-amino-2,3-dihydrobenzofuran, 2,3-dihydro-5-benzofuranamine, 2,3-dihydrobenzo b furan-5-amine, pubchem16462, 2,3-dihydro-5-benzofuranamine;, 2,3-dihydrobenzofuran-5-amine; |
PubChem CID | 3841102 |
IUPAC Name | 2,3-dihydro-1-benzofuran-5-amine |
SMILES | C1COC2=C1C=C(C=C2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426157
|
Thermo Scientific Alfa Aesar
H33344.MD |
250 mg |
57.70 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15406157
|
Thermo Scientific Alfa Aesar
H33344.03 |
1 g | N/A | ||||||
15416157
|
Thermo Scientific Alfa Aesar
H33344.06 |
5 g | N/A | ||||||
Description
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Chemical Identifiers
42933-43-7 | |
135.166 | |
YJMADHMYUJFMQE-UHFFFAOYSA-N | |
3841102 | |
C1COC2=C1C=C(C=C2)N |
C8H9NO | |
MFCD03617969 | |
2,3-dihydrobenzofuran-5-amine, 2,3-dihydrobenzo b furan-5-ylamine, 5-amino-2,3-dihydrobenzo b furan, 5-aminocoumaran, 5-amino-2,3-dihydrobenzofuran, 2,3-dihydro-5-benzofuranamine, 2,3-dihydrobenzo b furan-5-amine, pubchem16462, 2,3-dihydro-5-benzofuranamine;, 2,3-dihydrobenzofuran-5-amine; | |
2,3-dihydro-1-benzofuran-5-amine |
Specifications
42933-43-7 | |
C8H9NO | |
Air Sensitive | |
Sparingly soluble in water.(0.26g/L) (25°C), | |
C1COC2=C1C=C(C=C2)N | |
135.166 | |
135.17 | |
5-Amino-2,3-dihydrobenzo[b]furan |
78°C to 80°C | |
MFCD03617969 | |
2,3-dihydrobenzofuran-5-amine, 2,3-dihydrobenzo b furan-5-ylamine, 5-amino-2,3-dihydrobenzo b furan, 5-aminocoumaran, 5-amino-2,3-dihydrobenzofuran, 2,3-dihydro-5-benzofuranamine, 2,3-dihydrobenzo b furan-5-amine, pubchem16462, 2,3-dihydro-5-benzofuranamine;, 2,3-dihydrobenzofuran-5-amine; | |
YJMADHMYUJFMQE-UHFFFAOYSA-N | |
2,3-dihydro-1-benzofuran-5-amine | |
3841102 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.