Learn More
5-Amino-2,3-dihydrobenzofuran, 97%, Thermo Scientific Chemicals
136.00 € - 234.00 €
Chemical Identifiers
CAS | 42933-43-7 |
---|---|
Molecular Formula | C8H9NO |
Molecular Weight (g/mol) | 135.17 |
InChI Key | YJMADHMYUJFMQE-UHFFFAOYSA-N |
Synonym | 2,3-dihydrobenzofuran-5-amine, 2,3-dihydrobenzo b furan-5-ylamine, 5-amino-2,3-dihydrobenzo b furan, 5-aminocoumaran, 5-amino-2,3-dihydrobenzofuran, 2,3-dihydro-5-benzofuranamine, 2,3-dihydrobenzo b furan-5-amine, pubchem16462, 2,3-dihydro-5-benzofuranamine;, 2,3-dihydrobenzofuran-5-amine; |
PubChem CID | 3841102 |
IUPAC Name | 2,3-dihydro-1-benzofuran-5-amine |
SMILES | C1COC2=C1C=C(C=C2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10785072
|
Thermo Scientific Acros
438030010 |
1 g |
136.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10891691
|
Thermo Scientific Acros
438030050 |
5 g |
234.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
42933-43-7 | |
135.17 | |
2,3-dihydrobenzofuran-5-amine, 2,3-dihydrobenzo b furan-5-ylamine, 5-amino-2,3-dihydrobenzo b furan, 5-aminocoumaran, 5-amino-2,3-dihydrobenzofuran, 2,3-dihydro-5-benzofuranamine, 2,3-dihydrobenzo b furan-5-amine, pubchem16462, 2,3-dihydro-5-benzofuranamine;, 2,3-dihydrobenzofuran-5-amine; | |
2,3-dihydro-1-benzofuran-5-amine |
C8H9NO | |
YJMADHMYUJFMQE-UHFFFAOYSA-N | |
3841102 | |
C1COC2=C1C=C(C=C2)N |
Specifications
42933-43-7 | |
100.0 | |
White to Brown | |
96% min. (GC) | |
C8H9NO | |
YJMADHMYUJFMQE-UHFFFAOYSA-N | |
2,3-dihydro-1-benzofuran-5-amine | |
3841102 | |
97% | |
5-Amino-2, 3-dihydrobenzofuran |
96.0 | |
81.6°C to 83.4°C | |
Authentic | |
Glass bottle | |
2,3-dihydrobenzofuran-5-amine, 2,3-dihydrobenzo b furan-5-ylamine, 5-amino-2,3-dihydrobenzo b furan, 5-aminocoumaran, 5-amino-2,3-dihydrobenzofuran, 2,3-dihydro-5-benzofuranamine, 2,3-dihydrobenzo b furan-5-amine, pubchem16462, 2,3-dihydro-5-benzofuranamine;, 2,3-dihydrobenzofuran-5-amine; | |
C1COC2=C1C=C(C=C2)N | |
135.17 | |
135.17 | |
Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Co
GHS Signal Word: Warning
- 5-Amino-2,3-dihydrobenzofuran
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.