missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-fluorophenol, 97%, Thermo Scientific™

88.49 € - 213.00 €

Chemical Identifiers

CAS 100367-48-4
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD13185335
InChI Key XQYICHLOAMMXKE-UHFFFAOYSA-N
Synonym phenol, 5-amino-2-fluoro, 4-fluoro-3-hydroxyaniline, aminofluorophenol, acmc-20e9pp, 3-amino-6-fluorophenol, 4-fluoro-3-hydroxy-aniline
PubChem CID 22641413
IUPAC Name 5-amino-2-fluorophenol
SMILES C1=CC(=C(C=C1N)O)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15466687
View Documents
Thermo Scientific Alfa Aesar
H33821.MD
250 mg
88.49 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15456687
View Documents
Thermo Scientific Alfa Aesar
H33821.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Amino-2-fluorophenol is used as synthetic building block.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

100367-48-4
127.118
XQYICHLOAMMXKE-UHFFFAOYSA-N
22641413
C1=CC(=C(C=C1N)O)F
C6H6FNO
MFCD13185335
phenol, 5-amino-2-fluoro, 4-fluoro-3-hydroxyaniline, aminofluorophenol, acmc-20e9pp, 3-amino-6-fluorophenol, 4-fluoro-3-hydroxy-aniline
5-amino-2-fluorophenol

Specifications

100367-48-4
White to Yellow
MFCD13185335
XQYICHLOAMMXKE-UHFFFAOYSA-N
5-amino-2-fluorophenol
22641413
97%
5-Amino-2-fluorophenol
156°C to 158°C
C6H6FNO
phenol, 5-amino-2-fluoro, 4-fluoro-3-hydroxyaniline, aminofluorophenol, acmc-20e9pp, 3-amino-6-fluorophenol, 4-fluoro-3-hydroxy-aniline
C1=CC(=C(C=C1N)O)F
127.118
127.12
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.