missing translation for 'onlineSavingsMsg'
Learn More
5-Amino-4H-1,2,4-triazole-3-thiol, tech., Thermo Scientific™ Product Code.: 10669724

5-Amino-4H-1,2,4-triazole-3-thiol, tech., Thermo Scientific™

Product Code. 10669724
250 mg
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy

Product Code. 10669724

Brand: Thermo Scientific Maybridge SB00513CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16691-43-3
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.14
MDL Number MFCD00005231
InChI Key WZUUZPAYWFIBDF-UHFFFAOYSA-N
Synonym 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
PubChem CID 2723869
IUPAC Name 5-amino-1,2-dihydro-1,2,4-triazole-3-thione
SMILES NC1=NC(=S)NN1

Specifications

Quantity 250 mg
Formula Weight 116.15
Chemical Name or Material 5-Amino-4H-1, 2, 4-triazole-3-thiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Amino-4H-1,2,4-triazole-3-thiol, tech., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.