missing translation for 'onlineSavingsMsg'
Learn More

5-Aminobenzene-1,3-diol hydrochloride, Technical Grade, Thermo Scientific™

108.00 € - 884.00 €

Chemical Identifiers

CAS 6318-56-5
Molecular Formula C6H8ClNO2
Molecular Weight (g/mol) 161.59
MDL Number MFCD00662897
InChI Key VNZZCDQPCQIUGG-UHFFFAOYSA-N
Synonym 5-aminobenzene-1,3-diol hydrochloride, 1,3-benzenediol, 5-amino-, hydrochloride, 5-aminoresorcinol, hydrochloride, pubchem19962, 5-aminobenzene-1,3-diol hcl, 5-aminobenzene-1,3-diol hydrochloridyl
PubChem CID 459248
SMILES Cl.NC1=CC(O)=CC(O)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10613203
View Documents
Thermo Scientific Maybridge
BTB10193CB
250 mg
108.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10446262
View Documents
Thermo Scientific Maybridge
BTB10193DA
1 g
217.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10274684
View Documents
Thermo Scientific Maybridge
BTB10193DE
5 g
884.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

6318-56-5
161.59
VNZZCDQPCQIUGG-UHFFFAOYSA-N
459248
C6H8ClNO2
MFCD00662897
5-aminobenzene-1,3-diol hydrochloride, 1,3-benzenediol, 5-amino-, hydrochloride, 5-aminoresorcinol, hydrochloride, pubchem19962, 5-aminobenzene-1,3-diol hcl, 5-aminobenzene-1,3-diol hydrochloridyl
Cl.NC1=CC(O)=CC(O)=C1

Specifications

6318-56-5
NH2C6H3(OH)2.HCl
5-aminobenzene-1,3-diol hydrochloride, 1,3-benzenediol, 5-amino-, hydrochloride, 5-aminoresorcinol, hydrochloride, pubchem19962, 5-aminobenzene-1,3-diol hcl, 5-aminobenzene-1,3-diol hydrochloridyl
Cl.NC1=CC(O)=CC(O)=C1
161.59
161.59
Technical
C6H8ClNO2
MFCD00662897
VNZZCDQPCQIUGG-UHFFFAOYSA-N
5-aminobenzene-1,3-diol;hydrochloride
459248
Tech.
5-Aminobenzene-1, 3-diol hydrochloride
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.