missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminoindan-1-one, 97%, Thermo Scientific™
179.00 € - 909.00 €
Chemical Identifiers
CAS | 3470-54-0 |
---|---|
Molecular Formula | C9H9NO |
Molecular Weight (g/mol) | 147.177 |
InChI Key | HODOSJNSRPXYBH-UHFFFAOYSA-N |
Synonym | 5-aminoindan-1-one, 5-amino-1-indanone, 5-amino-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-amino-2,3-dihydro, 5-aminoindanone, 5-amino-1-oxoindane, 5-amino-indan-1-one, pubchem14476, 5-amino-1-indanone #, 5-azanyl-2,3-dihydroinden-1-one |
PubChem CID | 312890 |
IUPAC Name | 5-amino-2,3-dihydroinden-1-one |
SMILES | C1CC(=O)C2=C1C=C(C=C2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10099411
|
Thermo Scientific Maybridge
NRB01003DA |
1 g |
179.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10650866
|
Thermo Scientific Maybridge
NRB01003DE |
5 g |
587.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10081964
|
Thermo Scientific Maybridge
NRB01003EA |
10 g |
909.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
3470-54-0 | |
147.177 | |
5-aminoindan-1-one, 5-amino-1-indanone, 5-amino-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-amino-2,3-dihydro, 5-aminoindanone, 5-amino-1-oxoindane, 5-amino-indan-1-one, pubchem14476, 5-amino-1-indanone #, 5-azanyl-2,3-dihydroinden-1-one | |
5-amino-2,3-dihydroinden-1-one |
C9H9NO | |
HODOSJNSRPXYBH-UHFFFAOYSA-N | |
312890 | |
C1CC(=O)C2=C1C=C(C=C2)N |
Specifications
3470-54-0 | |
Amber glass bottle | |
5-aminoindan-1-one, 5-amino-1-indanone, 5-amino-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-amino-2,3-dihydro, 5-aminoindanone, 5-amino-1-oxoindane, 5-amino-indan-1-one, pubchem14476, 5-amino-1-indanone #, 5-azanyl-2,3-dihydroinden-1-one | |
C1CC(=O)C2=C1C=C(C=C2)N | |
147.177 | |
147.18 | |
5-Aminoindan-1-one |
97% | |
C9H9NO | |
HODOSJNSRPXYBH-UHFFFAOYSA-N | |
5-amino-2,3-dihydroinden-1-one | |
312890 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title