missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals

Product Code. 11477367
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Aminoisophthalic acid is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminoisophthalic acid is a useful biochemical for proteomics research.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-31-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00002522
InChI Key KBZFDRWPMZESDI-UHFFFAOYSA-N
Synonym 5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
PubChem CID 66833
IUPAC Name 5-aminobenzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)N)C(=O)O

Specifications

Melting Point >300°C
Quantity 50 g
Beilstein 2805628
Solubility Information Insoluble in water.
Formula Weight 181.15
Percent Purity 95%
Chemical Name or Material 5-Aminoisophthalic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'