missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Aminosulfonyl)-1-methyl-1H-pyrrole-2-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Specifications
Specifications
CAS | 306936-62-9 |
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C6H8N2O4S |
MDL Number | MFCD02677728 |
Quantity | 1 g |
Synonym | 5-aminosulfonyl-1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-5-sulfamoyl-1h-pyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylicacid, 5-aminosulfonyl-1-methyl, 5-aminosulphonyl-1-methyl-1h-pyrrole-2-carboxylic acid |
InChI Key | HIUOQXPHVZAOBG-UHFFFAOYSA-N |
SMILES | CN1C(=CC=C1S(=O)(=O)N)C(=O)O |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction