missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1,3,4-thiadiazol-2-ylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 180.03 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-1, 3, 4-thiadiazol-2-ylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'