missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-benzofuran, 97%, Thermo Scientific™

Product Code. p-2701270
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10641673

Brand: Thermo Scientific Maybridge CC07110CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 23145-07-5
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
MDL Number MFCD03407317
InChI Key AYOVPQORFBWFNO-UHFFFAOYSA-N
Synonym 5-bromobenzofuran, benzofuran, 5-bromo, 5-bromobenzo b furan, 5-bromo-benzofuran, 5-bromobenzo beta furan, buttpark 95\50-66, pubchem22017, 5-bromobenzo b-furan, #
PubChem CID 90015
IUPAC Name 5-bromo-1-benzofuran
SMILES C1=CC2=C(C=CO2)C=C1Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-benzofuran
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.