missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-bromo-1-methyl-1H-indole-3-carbaldehyde, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 238.08 |
Percent Purity | 97% |
Chemical Name or Material | 5-bromo-1-methyl-1H-indole-3-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'