missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methylimidazole, 97%, Thermo Scientific Chemicals

Product Code. 10794955
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-21-0
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.00
MDL Number MFCD01632218
InChI Key HATLLUIOEIXWGD-UHFFFAOYSA-N
Synonym 5-bromo-1-methyl-1h-imidazole, 5-bromo-n-methylimidazole, pubchem8951, 1h-imidazole, 5-bromo-1-methyl, 5-bromo-1-methyl-imidazole, imidazole,5-bromo-1-methyl, 5-bromo-n-methyl-imidazole, 5-bromo-1-methyl imidazole, acmc-1c6k3, 1-methyl-5-bromo-1h-imidazole
PubChem CID 1515258
IUPAC Name 5-bromo-1-methylimidazole
SMILES CN1C=NC=C1Br

Specifications

Melting Point 40°C to 41°C
Boiling Point 120°C (15.0 mmHg)
Flash Point 82°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 161
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-methylimidazole
compliance-icons compliance-icons
Product Identifier
  • 5-Bromo-1-methylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'