missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,1,3-benzoxadiazole, 97%, Thermo Scientific™

Product Code. 10384322
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51376-06-8
Molecular Formula C6H3BrN2O
Molecular Weight (g/mol) 199.01
MDL Number MFCD02682026
InChI Key ZWDFFESFCIACQC-UHFFFAOYSA-N
Synonym 5-bromobenzo c 1,2,5 oxadiazole, 5-bromobenzofurazan, 2,1,3-benzoxadiazole, 5-bromo, 5-bromo-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole,5-bromo, 5-bromobenzfurazan, pubchem15922, acmc-209ktf, amth003, buttpark 85\04-54
PubChem CID 2776298
IUPAC Name 5-bromo-2,1,3-benzoxadiazole
SMILES BrC1=CC2=NON=C2C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.01
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 1, 3-benzoxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'