missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-nitrophenol, 96%, Thermo Scientific Chemicals

Product Code. 15472399
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in ethyl acetate and methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27684-84-0
Molecular Formula C6H4BrNO3
Molecular Weight (g/mol) 218.006
MDL Number MFCD03095027
InChI Key DTWHNSNSUBKGTC-UHFFFAOYSA-N
Synonym 5-bromo-2-nitrophenol, phenol, 5-bromo-2-nitro, 3-bromo-6-nitrophenol, 4-bromo-2-hydroxynitrobenzene, 2-nitro-5-bromophenol, 5-bromo-2-nitro-phenol, phenol,5-bromo-2-nitro, pubchem16073, acmc-1cct0, 5-bromanyl-2-nitro-phenol
PubChem CID 13970496
SMILES C1=CC(=C(C=C1Br)O)[N+](=O)[O-]

Specifications

Melting Point 40°C to 41°C
Color Yellow
Quantity 5 g
Beilstein 2329085
Solubility Information Soluble in ethyl acetate and methanol.
Formula Weight 218.01
Percent Purity 96%
Physical Form Crystalline Solid
Chemical Name or Material 5-Bromo-2-nitrophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'