missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-pyridineacetic acid, 98+%, Thermo Scientific Chemicals

116.00 € - 433.00 €

Chemical Identifiers

CAS 39891-12-8
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00829308
InChI Key UETIDNDXXGCJCE-UHFFFAOYSA-N
Synonym 5-bromo-3-pyridylacetic acid, 2-5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridineacetic acid, 3-pyridineacetic acid, 5-bromo, 2-5-bromo-3-pyridyl acetic acid, 5-bromo-3-pyridylaceticacid, 5-bromo-pyridin-3-yl-acetic acid, 5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridinyl acetic acid, 2-5-bromopyridin-3-yl acetic acid
PubChem CID 2802539
IUPAC Name 2-(5-bromopyridin-3-yl)acetic acid
SMILES C1=C(C=NC=C1Br)CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393147
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13579.03
1 g
116.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11303157
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13579.06
5 g
433.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

39891-12-8
216.034
UETIDNDXXGCJCE-UHFFFAOYSA-N
2802539
C1=C(C=NC=C1Br)CC(=O)O
C7H6BrNO2
MFCD00829308
5-bromo-3-pyridylacetic acid, 2-5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridineacetic acid, 3-pyridineacetic acid, 5-bromo, 2-5-bromo-3-pyridyl acetic acid, 5-bromo-3-pyridylaceticacid, 5-bromo-pyridin-3-yl-acetic acid, 5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridinyl acetic acid, 2-5-bromopyridin-3-yl acetic acid
2-(5-bromopyridin-3-yl)acetic acid

Specifications

39891-12-8
C7H6BrNO2
388264
Soluble in water.
C1=C(C=NC=C1Br)CC(=O)O
216.034
216.03
5-Bromo-3-pyridineacetic acid
182°C to 185°C
MFCD00829308
5-bromo-3-pyridylacetic acid, 2-5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridineacetic acid, 3-pyridineacetic acid, 5-bromo, 2-5-bromo-3-pyridyl acetic acid, 5-bromo-3-pyridylaceticacid, 5-bromo-pyridin-3-yl-acetic acid, 5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridinyl acetic acid, 2-5-bromopyridin-3-yl acetic acid
UETIDNDXXGCJCE-UHFFFAOYSA-N
2-(5-bromopyridin-3-yl)acetic acid
2802539
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only