missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, MP Biomedicals™

Product Code. 11406402
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Commonly known as X-Gal. 5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, MP Biomedicals is a histochemical substrate for β-galactosidase. It is particularly useful in molecular biology, where β-galactosidase is frequently used as a report

5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside is used as indigogenic substrate for β-galactosidase, for detection of β-galactosidase-positive clones, and the identification of lac and bacterial colonies or phage plaques.

It is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

  • Insertion of DNA into the lac Z region results in the loss of β-galactosidase activity
  • Lac+ bacterial colonies resulting from a-complementation will appear blue
  • Bacterial colonies containing plasmid with DNA inserted in the lac Z region will be incapable of a-complementation and will appear white
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Melting Point 230°C (decomposition)(literature)
Quantity 1 g
Solubility Information Soluble in DMF (2% w/v, clear and colorless solution).
Formula Weight 408.6
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.