missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-6-chloronicotinic acid, 97%, Thermo Scientific Chemicals

Product Code. 15435107
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29241-62-1
Molecular Formula C6H2BrClNO2
Molecular Weight (g/mol) 235.44
MDL Number MFCD01927098
InChI Key DXEUARPQHJXMII-UHFFFAOYSA-M
Synonym 5-bromo-6-chloronicotinic acid, 5-bromo-6-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 5-bromo-6-chloro, 3-bromo-2-chloropyridine-5-carboxylic acid, 3-bromo-2-chloro-5-pyridinecarboxylic acid, zlchem 1120, pubchem12922, acmc-1ckhn, ksc203c5j, abbypharma ap-12-5123
PubChem CID 285433
SMILES [O-]C(=O)C1=CN=C(Cl)C(Br)=C1

Specifications

Melting Point 170°C to 173°C
Quantity 1 g
IUPAC Name 5-bromo-6-chloropyridine-3-carboxylate
Formula Weight 236.45
Percent Purity 97%
Chemical Name or Material 5-Bromo-6-chloronicotinic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'