missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]thiophene, 98+%, Thermo Scientific Chemicals

Product Code. 11354586
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4923-87-9
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.09
MDL Number MFCD03069318
InChI Key RDSIMGKJEYNNLF-UHFFFAOYSA-N
Synonym 5-bromobenzo b thiophene, 5-bromobenzothiophene, 5-bromo-benzo b thiophene, 5-bromothianaphthene, benzo b thiophene, 5-bromo, zlchem 181, pubchem7752, 5-bromo-benzothiophene, pubchem15353, acmc-1anox
PubChem CID 2776578
IUPAC Name 5-bromo-1-benzothiophene
SMILES BrC1=CC=C2SC=CC2=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 105°C to 107°C (10 mmHg)
Quantity 5 g
Beilstein 112878
Sensitivity Light sensitive
Formula Weight 213.1
Percent Purity ≥98%
Chemical Name or Material 5-Bromobenzo[b]thiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'