missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzofurazan, 97%, Thermo Scientific Chemicals

Product Code. 15448137
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51376-06-8
Molecular Formula C6H3BrN2O
MDL Number MFCD02682026
Quantity 1 g
Synonym 5-bromobenzo c 1,2,5 oxadiazole, 5-bromobenzofurazan, 2,1,3-benzoxadiazole, 5-bromo, 5-bromo-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole,5-bromo, 5-bromobenzfurazan, pubchem15922, acmc-209ktf, amth003, buttpark 85\04-54
InChI Key ZWDFFESFCIACQC-UHFFFAOYSA-N
SMILES BrC1=CC2=NON=C2C=C1
IUPAC Name 5-bromo-2,1,3-benzoxadiazole
Molecular Weight (g/mol) 199.01
PubChem CID 2776298
Formula Weight 199.01
Percent Purity 97%
Chemical Name or Material 5-Bromobenzofurazan
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'