missing translation for 'onlineSavingsMsg'
Learn More
5-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific Chemicals Product Code.: 15496047

5-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific Chemicals

Product Code. 15496047
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 15496047

Brand: Thermo Scientific Alfa Aesar H33240.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69214-09-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD08275698
InChI Key CCOFGVWHMYYDBG-UHFFFAOYSA-N
Synonym 5-bromoimidazo 1,2-a pyridine, 5-bromo-imidazo 1,2-a pyridine, 5-bromoh-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 5-bromo, acmc-209o6n, 5-bromo-4-hydroimidazo 1,2-a pyridineShow MoreShow Less
PubChem CID 13037241
IUPAC Name 5-bromoimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=CN2C(=C1)Br

Specifications

Melting Point 61°C to 64°C
Quantity 1 g
Formula Weight 197.03
Percent Purity ≥97%
Chemical Name or Material 5-Bromoimidazo[1,2-a]pyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10178607 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog NumberH33240.03,H33240.09
Lot Number10178607 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33240.03,H33240.09
Lot Number10170030 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33240.03,H33240.09
Lot Number10227050 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33240.03,H33240.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.