missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-3-carboxaldehyde, 98+%, Thermo Scientific™

Product Code. 15409089
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 877-03-2
Melting Point 204°C to 206°C
Molecular Formula C9H6BrNO
MDL Number MFCD00152016
Quantity 1 g
Beilstein 124725
Synonym 5-bromoindole-3-carboxaldehyde, 5-bromoindole-3-carbaldehyde, 5-bromoindole-3-carboxyaldehyde, 5-bromo-3-formylindole, 5-bromoindole-3-aldehyde, 5-bromo-3-formyl-1h-indole, 5-bromo-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 5-bromo, pubchem9122, acmc-209qpj
InChI Key PEENKJZANBYXNB-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1Br)C(=CN2)C=O
IUPAC Name 5-bromo-1H-indole-3-carbaldehyde
Molecular Weight (g/mol) 224.057
PubChem CID 70137
Formula Weight 224.06
Percent Purity ≥98%
Chemical Name or Material 5-Bromoindole-3-carboxaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'