Learn More
5-Bromoisatin, 98%, Thermo Scientific Chemicals
27.44 € - 88.24 €
Chemical Identifiers
CAS | 87-48-9 |
---|---|
Molecular Formula | C8H4BrNO2 |
Molecular Weight (g/mol) | 226.03 |
MDL Number | MFCD00005719 |
InChI Key | MBVCESWADCIXJN-UHFFFAOYSA-N |
Synonym | 5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate |
PubChem CID | 6889 |
IUPAC Name | 5-bromo-1H-indole-2,3-dione |
SMILES | C1=CC2=C(C=C1Br)C(=O)C(=O)N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10126433
|
Thermo Scientific Acros
154230050 |
5 g | Glass bottle |
27.44 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10538873
|
Thermo Scientific Acros
154230250 |
25 g | Plastic Bottle |
88.24 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
87-48-9 | |
226.03 | |
MBVCESWADCIXJN-UHFFFAOYSA-N | |
6889 | |
C1=CC2=C(C=C1Br)C(=O)C(=O)N2 |
C8H4BrNO2 | |
MFCD00005719 | |
5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate | |
5-bromo-1H-indole-2,3-dione |
Specifications
87-48-9 | |
100.0 | |
Orange | |
97.5% min. (HPLC) | |
MFCD00005719 | |
5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate | |
C1=CC2=C(C=C1Br)C(=O)C(=O)N2 | |
226.03 | |
226.03 | |
Crystalline Powder |
97.5 | |
251.0°C to 253.0°C | |
Authentic | |
C8H4BrNO2 | |
21, 453 | |
MBVCESWADCIXJN-UHFFFAOYSA-N | |
5-bromo-1H-indole-2,3-dione | |
6889 | |
98% | |
5-Bromoisatin |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-747-7
- 5-Bromoisatin
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.