missing translation for 'onlineSavingsMsg'
Learn More
5-(Bromomethyl)-2,1,3-benzoxadiazole, 97%, Thermo Scientific™ Product Code.: 10765915

5-(Bromomethyl)-2,1,3-benzoxadiazole, 97%, Thermo Scientific™

Product Code. 10765915
10 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10765915

Brand: Thermo Scientific Maybridge CC09708EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 32863-31-3
Molecular Formula C7H5BrN2O
Molecular Weight (g/mol) 213.03
MDL Number MFCD00174369
InChI Key BUJFLTNYWUEROF-UHFFFAOYSA-N
Synonym 5-bromomethyl-2,1,3-benzoxadiazole, 5-bromomethyl benzo c 1,2,5 oxadiazole, 5-bromomethylbenzofurazan, 5-bromomethyl benzofurazan, 2,1,3-benzoxadiazole, 5-bromomethyl, zlchem 1034, 5-bromomethylbenzofurazane, 5-bromomethyl benzo c furazan, 5-bromomethyl-benzo 1,2,5 oxadiazole, 5-bromomethyl-2,1,3-benzooxadiazoleShow MoreShow Less
PubChem CID 2735587
IUPAC Name 5-(bromomethyl)-2,1,3-benzoxadiazole
SMILES BrCC1=CC2=NON=C2C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 213.03
Percent Purity 97%
Chemical Name or Material 5-(Bromomethyl)-2, 1, 3-benzoxadiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(Bromomethyl)-2,1,3-benzoxadiazole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.