missing translation for 'onlineSavingsMsg'
Learn More

5-Bromophthalide, 98%

Product Code. p-7032434
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15456777

Brand: Thermo Scientific Alfa Aesar H33891.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64169-34-2 | C8H5BrO2 | 213.03 g/mol

5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.

Solubility
Soluble in acetone.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64169-34-2
Molecular Formula C8H5BrO2
Molecular Weight (g/mol) 213.03
MDL Number MFCD01797360
InChI Key IUSPXLCLQIZFHL-UHFFFAOYSA-N
Synonym 5-bromophthalide, 5-bromoisobenzofuran-1 3h-one, 5-bromo-1 3h-isobenzofuranone, 5-bromo-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 5-bromo, 5-bromo-2-benzofuran-1 3h-one, 5-bromo-3-hydroisobenzofuran-1-one, 5-bromo-1,3-dihydro-2-benzofuran-1-one, 5-bromo-1,3-dihydro-isobenzofuran-1-one, 5-bromphthalide
PubChem CID 603144
IUPAC Name 5-bromo-1,3-dihydro-2-benzofuran-1-one
SMILES BrC1=CC=C2C(=O)OCC2=C1

Specifications

Melting Point 162°C to 166°C
Quantity 5 g
Solubility Information Soluble in acetone.
Formula Weight 213.03
Percent Purity 98%
Chemical Name or Material 5-Bromophthalide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.