missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Bromopyrid-2-yl)methanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 188.02 |
Percent Purity | 97% |
Chemical Name or Material | (5-Bromopyrid-2-yl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'