Learn More
5-Bromothiazole, 98%, Thermo Scientific Chemicals
38.69 € - 303.00 €
Chemical Identifiers
CAS | 3034-55-7 |
---|---|
Molecular Formula | C3H2BrNS |
Molecular Weight (g/mol) | 164.02 |
MDL Number | MFCD07787394 |
InChI Key | DWUPYMSVAPQXMS-UHFFFAOYSA-N |
PubChem CID | 546059 |
IUPAC Name | 5-bromo-1,3-thiazole |
SMILES | C1=C(SC=N1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15461608
|
Thermo Scientific Alfa Aesar
H55387.MD |
250 mg |
38.69 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15441608
|
Thermo Scientific Alfa Aesar
H55387.03 |
1 g |
91.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15451608
|
Thermo Scientific Alfa Aesar
H55387.06 |
5 g |
303.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Bromothiazole is used as a pharmaceutical intermediate and performance material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromothiazole is used as a pharmaceutical intermediate and performance material.
Solubility
Soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
3034-55-7 | |
164.02 | |
DWUPYMSVAPQXMS-UHFFFAOYSA-N | |
5-bromo-1,3-thiazole |
C3H2BrNS | |
MFCD07787394 | |
546059 | |
C1=C(SC=N1)Br |
Specifications
3034-55-7 | |
76°C (169°F) | |
1.5955 | |
Moisture sensitive | |
Soluble in water. | |
C1=C(SC=N1)Br | |
164.02 | |
164.02 | |
5-Bromothiazole |
1.835 | |
C3H2BrNS | |
MFCD07787394 | |
5-bromothiazole, thiazole, 5-bromo, 5-bromo-thiazole, 5-bromthiazole, thiazole,5-bromo, 5-bromothiazol, 5-bromo thiazole, zlchem 497, pubchem2077, 2-bromo-1,4-thiazole | |
DWUPYMSVAPQXMS-UHFFFAOYSA-N | |
5-bromo-1,3-thiazole | |
546059 | |
98% |
Safety and Handling
GHS H Statement
H301-H318-H227-H315-H317-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H302-H315-H317-H318-H335
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only