missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxaldehyde, 95%, Thermo Scientific™

Product Code. 11305539
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27006-76-4
Molecular Formula C6H7ClN2O
Molecular Weight (g/mol) 158.585
MDL Number MFCD00052538
InChI Key SZRSMNYUEXXEBL-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole-4-carbaldehyde, 5-chloro-1,3-dimethyl-1h-pyrazole-4-carboxaldehyde, 5-chloro-1,3-dimethyl pyrazole-4-aldehyde, 5-chloro-1,3-dimethylpyrazole-4-carboxaldehyde, 5-chloro-4-formyl-1,3-dimethylpyrazole, 5-chloro-1,3-dimethyl-1h-pyrazole-4-aldehyde, 1h-pyrazole-4-carboxaldehyde, 5-chloro-1,3-dimethyl, pubchem23629, acmc-20a2nm, ksc496s9t
PubChem CID 2777397
IUPAC Name 5-chloro-1,3-dimethylpyrazole-4-carbaldehyde
SMILES CC1=NN(C(=C1C=O)Cl)C

Specifications

Quantity 5 g
Formula Weight 158.59
Percent Purity 95%
Chemical Name or Material 5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'