missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,3-dimethyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Product Code. 11459253
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54454-10-3
Molecular Formula C5H7ClN2
Molecular Weight (g/mol) 130.58
MDL Number MFCD00051650
InChI Key DDUSLFAWARYAPR-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole, 5-chloro-1,3-dimethyl, 1,3-dimethyl-5-chloropyrazole, pubchem9980, acmc-20am54, 5-chloro-1,3-dimethyl-pyrazole, 5-chloranyl-1,3-dimethyl-pyrazole, 1,3-dimethyl-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazole #
PubChem CID 521507
IUPAC Name 5-chloro-1,3-dimethylpyrazole
SMILES CN1N=C(C)C=C1Cl

Specifications

Density 1.137
Boiling Point 153°C to 155°C
Refractive Index 1.486
Quantity 5 g
Beilstein 113451
Formula Weight 130.58
Percent Purity 98%
Chemical Name or Material 5-Chloro-1,3-dimethyl-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'