missing translation for 'onlineSavingsMsg'
Learn More
5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™ Product Code.: 10511112

5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10511112
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10511112

Brand: Thermo Scientific Maybridge KM08674DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175203-78-8
Molecular Formula C6H2Cl2N2O3S
Molecular Weight (g/mol) 253.05
MDL Number MFCD00203878
InChI Key ZFBZICACMDZKNQ-UHFFFAOYSA-N
Synonym 5-chlorobenzofurazan-4-sulfonyl chloride, 5-chlorobenzo c 1,2,5 oxadiazole-4-sulfonyl chloride, pubchem5077, 2,1,3-benzoxadiazole-4-sulfonylchloride,5-chloro, 5-chloro-2,1,3-benzoxadiazole-4-sulphonyl chloride, 2,1,3-benzoxadiazole-4-sulfonylchloride, 5-chloro, 5-chloranyl-2,1,3-benzoxadiazole-4-sulfonyl chloride, 5-chloro-benzo 1,2,5 oxadiazole-4-sulfonyl chlorideShow MoreShow Less
PubChem CID 2779275
IUPAC Name 5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride
SMILES ClC1=C(C2=NON=C2C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.06
Percent Purity 97%
Chemical Name or Material 5-chloro-2, 1, 3-benzoxadiazole-4-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.