missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-(trifluoromethoxy)aniline, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS | 326-64-7 |
Molecular Formula | C7H5ClF3NO |
Synonym | 5-chloro-2-trifluoromethoxy aniline, 5-chloro-2-trifluoro methoxy aniline, 5-chloro-2-trifluoromethoxy benzenamine, benzenamine, 5-chloro-2-trifluoromethoxy, 5-chloro-2-trifluoromethoxy-phenylamine, 2-amino-4-chlorotrifluoro-methoxybenzene, 5-chloro-2-trifluoromethoxy phenylamine |
InChI Key | KSBUTBXMRFVLND-UHFFFAOYSA-N |
SMILES | C1=CC(=C(C=C1Cl)N)OC(F)(F)F |
IUPAC Name | 5-chloro-2-(trifluoromethoxy)aniline |
Molecular Weight (g/mol) | 211.568 |
PubChem CID | 10932748 |
Formula Weight | 211.57 |
Percent Purity | 97% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'