missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbonitrile, 99%, Thermo Scientific™

50.60 € - 213.00 €

Chemical Identifiers

CAS 51626-33-6
Molecular Formula C11H8ClN3
Molecular Weight (g/mol) 217.656
MDL Number MFCD00455809
InChI Key MWXQWRGAGVHIDR-UHFFFAOYSA-N
Synonym 5-chloro-3-methyl-1-phenyl-1h-pyrazole-4-carbonitrile, acmc-20am4a
PubChem CID 2748811
IUPAC Name 5-chloro-3-methyl-1-phenylpyrazole-4-carbonitrile
SMILES CC1=NN(C(=C1C#N)Cl)C2=CC=CC=C2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11490020
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50562.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11400030
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50562.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

51626-33-6
217.656
MWXQWRGAGVHIDR-UHFFFAOYSA-N
2748811
CC1=NN(C(=C1C#N)Cl)C2=CC=CC=C2
C11H8ClN3
MFCD00455809
5-chloro-3-methyl-1-phenyl-1h-pyrazole-4-carbonitrile, acmc-20am4a
5-chloro-3-methyl-1-phenylpyrazole-4-carbonitrile

Specifications

51626-33-6
MFCD00455809
5-chloro-3-methyl-1-phenyl-1h-pyrazole-4-carbonitrile, acmc-20am4a
CC1=NN(C(=C1C#N)Cl)C2=CC=CC=C2
217.656
217.66
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbonitrile
C11H8ClN3
UN3439
MWXQWRGAGVHIDR-UHFFFAOYSA-N
5-chloro-3-methyl-1-phenylpyrazole-4-carbonitrile
2748811
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications