missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™
17.70 € - 105.00 €
Chemical Identifiers
CAS | 166964-33-6 |
---|---|
Molecular Formula | C9H6Cl2O2S2 |
Molecular Weight (g/mol) | 281.17 |
MDL Number | MFCD00173797 |
InChI Key | JLTUANWNGKWRQO-UHFFFAOYSA-N |
Synonym | 5-chloro-3-methylbenzo b thiophene-2-sulfonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulphonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulfonyl, 5-chloro-3-methylbenzo b thiophene-2-sulphonylchloride, 5-chloro-3-methyl-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonylchloride, 5-chloro-3-methyl, zlchem 600, pubchem5100, buttpark 21\07-05, 5-chloro-3-methyl-benzothiophene-2-sulfonyl chloride |
PubChem CID | 2735756 |
IUPAC Name | 5-chloro-3-methyl-1-benzothiophene-2-sulfonyl chloride |
SMILES | CC1=C(SC2=CC=C(Cl)C=C12)S(Cl)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10580922
|
Thermo Scientific Maybridge
MWP00006DA |
1 g |
17.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396905
|
Thermo Scientific Maybridge
MWP00006EA |
10 g |
105.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
166964-33-6 | |
281.17 | |
JLTUANWNGKWRQO-UHFFFAOYSA-N | |
2735756 | |
CC1=C(SC2=CC=C(Cl)C=C12)S(Cl)(=O)=O |
C9H6Cl2O2S2 | |
MFCD00173797 | |
5-chloro-3-methylbenzo b thiophene-2-sulfonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulphonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulfonyl, 5-chloro-3-methylbenzo b thiophene-2-sulphonylchloride, 5-chloro-3-methyl-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonylchloride, 5-chloro-3-methyl, zlchem 600, pubchem5100, buttpark 21\07-05, 5-chloro-3-methyl-benzothiophene-2-sulfonyl chloride | |
5-chloro-3-methyl-1-benzothiophene-2-sulfonyl chloride |
Specifications
166964-33-6 | |
C9H6Cl2O2S2 | |
5-chloro-3-methylbenzo b thiophene-2-sulfonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulphonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulfonyl, 5-chloro-3-methylbenzo b thiophene-2-sulphonylchloride, 5-chloro-3-methyl-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonylchloride, 5-chloro-3-methyl, zlchem 600, pubchem5100, buttpark 21\07-05, 5-chloro-3-methyl-benzothiophene-2-sulfonyl chloride | |
CC1=C(SC2=CC=C(Cl)C=C12)S(Cl)(=O)=O | |
281.17 | |
281.18 | |
5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride |
97% | |
MFCD00173797 | |
JLTUANWNGKWRQO-UHFFFAOYSA-N | |
5-chloro-3-methyl-1-benzothiophene-2-sulfonyl chloride | |
2735756 | |
97% |